Learn More
Medchemexpress LLC HY-W018171 500mg Medchemexpress, 3,5,6-Trichloro-2-pyridinol CAS:6515-38-4 Purity:>98%
SDP
Catalog No. 502083417
Encompass_Preferred
Medchemexpress, HY-W018171 500mg 3,5,6-Trichloro-2-pyridinol CAS:6515-38-4 3,5,6-Trichloro-2-pyridinol (TCPy) is the main degradation product of the herbicide Triclopyr and the insecticides Chlorpyrifos and Chlorpyrifos-methyl. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3417 Supplier Medchemexpress LLC Supplier No. HYW018171500MG
Medchemexpress, HY-W018171 500mg 3,5,6-Trichloro-2-pyridinol CAS:6515-38-4 3,5,6-Trichloro-2-pyridinol (TCPy) is the main degradation product of the herbicide Triclopyr and the insecticides Chlorpyrifos and Chlorpyrifos-methyl. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.