Learn More
eMolecules​ 35661-60-0 | Fmoc-Leu-OH | Ambeed353.418 | C21H23NO4 | 98.000 | CC(C)C[C@H](NC(=O)OCC1c2ccccc2-c2ccccc12)C(O)=O | 25g | 552564879
Catalog No. 5000567290 Shop All eMolecules Inc. Products
Encompass_Preferred
Fmoc-Leu-OH | Ambeed | 35661-60-0353.418 | C21H23NO4 | 98.000 | CC(C)C[C@H](NC(=O)OCC1c2ccccc2-c2ccccc12)C(O)=O | 25g | 552564879
Catalog No. 50-005-67290 Supplier eMolecules​ Supplier No. 552564879
Fmoc-Leu-OH | Ambeed | 35661-60-0353.418 | C21H23NO4 | 98.000 | CC(C)C[C@H](NC(=O)OCC1c2ccccc2-c2ccccc12)C(O)=O | 25g | 552564879