Learn More
CEM Corporation 35 mL Pressure Vessels, Pyrex
SDP
Catalog No. 501877895
Encompass
The 35 mL pressure vessels have a working volume of 2.0-25.0 mL and are rated up to 300 psi.
Catalog No. 50-187-7895 Supplier CEM Corporation Supplier No. 909036
The 35 mL pressure vessels have a working volume of 2.0-25.0 mL and are rated up to 300 psi.