Learn More
Bellco Glass, Inc. 35mm Cell Culture Dish, with Gripping Ring, TC, Sterile, 20/pk, 500/cs
SDP
Catalog No. 502330529
Encompass_Preferred
35mm Cell Culture Dish, with Gripping Ring, TC, Sterile, 20/pk, 500/cs. cell culture dishes are ideal for all types of culturing where microscopic examination is required. Numeric indicators on the bottom of each plate allow users to identify the location of cells. • High clarity, 100% virgin polystyrene.• Flat transparent surface for distortion-free observation.• Vacuum plasma TC treatment, excellent cell adherence.• Stackable for easy storage and handling.• Sterilized by E-beam, SAL=10-6.• Non-Pyrogenic, DNase/Rnase free.
Catalog No. 50-233-0529 Supplier Bellco Glass, Inc. Supplier No. 451400058
35mm Cell Culture Dish, with Gripping Ring, TC, Sterile, 20/pk, 500/cs. cell culture dishes are ideal for all types of culturing where microscopic examination is required. Numeric indicators on the bottom of each plate allow users to identify the location of cells. • High clarity, 100% virgin polystyrene.• Flat transparent surface for distortion-free observation.• Vacuum plasma TC treatment, excellent cell adherence.• Stackable for easy storage and handling.• Sterilized by E-beam, SAL=10-6.• Non-Pyrogenic, DNase/Rnase free.