Learn More
Azzota Corp Cell Culture Dishes, Surface Treated, Dia. 35mm X 12mm (H), Polystyrene, Flat Bottom, Clear Color, Sterile, Package Of 20
Catalog No. 507532924
Encompass_Preferred
Cell Culture Dishes, Surface Treated, Dia. 35mm x 12mm (H), Polystyrene, Flat bottom, Clear color, Sterile, Package of 20
Catalog No. 50-753-2924 Supplier Azzota Corp Supplier No. PED35
Cell Culture Dishes, Surface Treated, Dia. 35mm x 12mm (H), Polystyrene, Flat bottom, Clear color, Sterile, Package of 20