Learn More
Cellvis 35 mm Glass bottom dish with 20 mm micro-well
Supplier:  Cellvis D35201.5H
Encompass
35mm glass bottom dish, dish size 35 mm, well size 20mm, #1.5 higher performance cover glass(0.170±0.005mm). Designed for high resolution imaging such as confocal microscopy. Recommended for Super Resolution Microscopy.
Catalog No. NC1757364