Learn More
Cellvis 35MM GLSBTM DSH 10MM MCRW#1.5
Catalog No. NC0589960
Encompass
35mm glass bottom dish, dish size 35mm, well size 10mm, #1.5 glass(0.16-0.19mm).
Catalog No. NC0589960 Supplier Cellvis Supplier No. D35101.5N
35mm glass bottom dish, dish size 35mm, well size 10mm, #1.5 glass(0.16-0.19mm).