Learn More
Ace Glass, Inc. 35MM GRADED SEALS 35MM GRADED SEALS
SDP
Catalog No. 50974605
Encompass
35mm quartz graded seals for connecting Pyrex or Kimax borosilicate glass to Quartz glass. Quartz length is 7.6cm, and borosilicate length is 10.2cm.
Catalog No. 50-974-605 Supplier Ace Glass, Inc. Supplier No. 845544
35mm quartz graded seals for connecting Pyrex or Kimax borosilicate glass to Quartz glass. Quartz length is 7.6cm, and borosilicate length is 10.2cm.