Learn More
Alkali Scientific ASI™ 35mm Nylon Multi Insert Dish, Sterile, Eazy Grip, 18/Cs, Groove, TC Treated
SDP
Catalog No. 503026810
Encompass_Preferred
Our new Multi Insert Dish with five small culture inserts assembled in one single dish. Multi Insert Dish is designed to study intercellular or inter-tissue interactions of more than two different cell or tissue species, successfully applicable to researches in biosystem mimicking in vivo condition. There are various types of dish available to choose depending on the intended use of the researchers. ▪ Mesh material: Nylon▪ Mesh thickness: 10 ~ 23 μm▪ Mesh pore size: 23 µm▪ Membrane material: Polycarbonate (PC)▪ Membrane thickness: 10 ~ 23 μm▪ Membrane pore size: 0.40 μm 
Catalog No. 50-302-6810 Supplier Alkali Scientific Supplier No. MDI0625
Our new Multi Insert Dish with five small culture inserts assembled in one single dish. Multi Insert Dish is designed to study intercellular or inter-tissue interactions of more than two different cell or tissue species, successfully applicable to researches in biosystem mimicking in vivo condition. There are various types of dish available to choose depending on the intended use of the researchers. ▪ Mesh material: Nylon▪ Mesh thickness: 10 ~ 23 μm▪ Mesh pore size: 23 µm▪ Membrane material: Polycarbonate (PC)▪ Membrane thickness: 10 ~ 23 μm▪ Membrane pore size: 0.40 μm 