Learn More
Alkali Scientific ASI™ 35mm PC Multi Insert Dish, Sterile, Eazy Grip, Non-Treated,18/Cs
SDP
Catalog No. 503028824
Encompass_Preferred
Our new Multi Insert Dish with five small culture inserts assembled in one single dish. Multi Insert Dish is designed to study intercellular or inter-tissue interactions of more than two different cell or tissue species, successfully applicable to researches in biosystem mimicking in vivo condition. There are various types of dish available to choose depending on the intended use of the researchers. • For the analysis of interactions between different cell types or tissues in a single dish by growing them in several inserts• Enables reproduction of the in vivo environment and organic system• Not treated• Sterile• External grip ring• 2 types of membranes:- nylon (thickness: 10 - 23 µm; porosity: 23 µm)- polycarbonate (thickness: 10 - 23 µm; porosity: 40 µm)
Catalog No. 50-302-8824 Supplier Alkali Scientific Supplier No. MDI0606
Our new Multi Insert Dish with five small culture inserts assembled in one single dish. Multi Insert Dish is designed to study intercellular or inter-tissue interactions of more than two different cell or tissue species, successfully applicable to researches in biosystem mimicking in vivo condition. There are various types of dish available to choose depending on the intended use of the researchers. • For the analysis of interactions between different cell types or tissues in a single dish by growing them in several inserts• Enables reproduction of the in vivo environment and organic system• Not treated• Sterile• External grip ring• 2 types of membranes:- nylon (thickness: 10 - 23 µm; porosity: 23 µm)- polycarbonate (thickness: 10 - 23 µm; porosity: 40 µm)