Learn More
CELLTREAT Scientific Products 35mm x 10mm Petri Dish, Sterile
SDP
Catalog No. 50202036
Encompass_Preferred
CELLTREAT Scientific Products 35mm x 10mm Non-Treated Petri Dish. Dishes have thicker and more durable plastic than other Petri Dishes. Vented lids contain molded spacers to allow gas exchange. Working volume of 5mL and growth area of 8.5cm¿. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile, 10/re-sealable bag, 960/case.
Catalog No. 50-202-036 Supplier CELLTREAT Scientific Products Supplier No. 229638
CELLTREAT Scientific Products 35mm x 10mm Non-Treated Petri Dish. Dishes have thicker and more durable plastic than other Petri Dishes. Vented lids contain molded spacers to allow gas exchange. Working volume of 5mL and growth area of 8.5cm¿. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile, 10/re-sealable bag, 960/case.