Learn More
Bellco Glass, Inc. 35mm x 10mm Petri Dish
SDP
Catalog No. 502329326
Encompass_Preferred
Cell Culture Dishes are available in Tissue Culture Treated and Nontreated Cell Culture versions. Petri Nontreated Cell Culture Dishes have thicker and more durable plastic than standard Petri Dishes – both can be used for the same applications. 60 70 and 100mm dishes available with a pronounced grip ring for improved handling.Specifications Vented lids contain molded spacers to allow gas exchange except Part No. 229655000 which has a nonvented lid With griping feature Working Volume 5mL Working Area 8.5cm2 Case Quantity 960
Catalog No. 50-232-9326 Supplier Bellco Glass, Inc. Supplier No. 229638000
Cell Culture Dishes are available in Tissue Culture Treated and Nontreated Cell Culture versions. Petri Nontreated Cell Culture Dishes have thicker and more durable plastic than standard Petri Dishes – both can be used for the same applications. 60 70 and 100mm dishes available with a pronounced grip ring for improved handling.Specifications Vented lids contain molded spacers to allow gas exchange except Part No. 229655000 which has a nonvented lid With griping feature Working Volume 5mL Working Area 8.5cm2 Case Quantity 960