Learn More
Bellco Glass, Inc. 35mm x 10mm Tissue Culture Treated Dish, Sterile
SDP
Supplier:  Bellco Glass, Inc. 229635000
Encompass_Preferred
Cell Culture Dishes have thicker and more durable plastic than Petri DishesVented lids contain molded spacers to allow gas exchange Pack Description10/Re-sealable BagCase Quantity500SterileYesPrimary ColorClearNon-pyrogenicYesPrimary MaterialPolystyreneWorking Volume5mLTissue Culture TreatedYesCell Growth Area8.5cm²RNase/DNase FreeYes
Catalog No. 50-232-9746