Learn More
Alkali Scientific 35μL GENEFECT™ Transfection Reagent
SDP
Catalog No. 502389027
Encompass_Preferred
GENEFECT™ is a new generation cationic polymer gene transfection reagent. It has several unique features necessary for efficient transfection, such as DNA condensation and endosomal release, which can improve gene transfection efficiency. Compared with cationic lipids, cationic polymers are very stable, easy to handle and more resistant to serum in cell culture. These advantages make gene transfection much easier and reproducible. GeneFect are widely used for both primary cell and established cell lines. Ideal for transient and stable transfection. GENEFECT™ Transfection Reagent prodcues a gentle transfection method for problematic cell types including primary cells. GENEFECT™ produces higher transfection efficiencies across a wide range of cell types and increased gene expression with very low toxicity.
Catalog No. 50-238-9027 Supplier Alkali Scientific Supplier No. GT360
GENEFECT™ is a new generation cationic polymer gene transfection reagent. It has several unique features necessary for efficient transfection, such as DNA condensation and endosomal release, which can improve gene transfection efficiency. Compared with cationic lipids, cationic polymers are very stable, easy to handle and more resistant to serum in cell culture. These advantages make gene transfection much easier and reproducible. GeneFect are widely used for both primary cell and established cell lines. Ideal for transient and stable transfection. GENEFECT™ Transfection Reagent prodcues a gentle transfection method for problematic cell types including primary cells. GENEFECT™ produces higher transfection efficiencies across a wide range of cell types and increased gene expression with very low toxicity.