Learn More
Chemglass Life Sciences Strainer Cap Only for FlowTubes, with 35µm Strainer Mesh, Sterile, 20 bags of 25 Caps
SDP
Catalog No. 501979487
Encompass_Preferred
Strainer Cap Only for FlowTubes, with 35¿m Strainer Mesh, Sterile, 20 bags of 25 Caps. Cap contains a 35¿m nylon mesh screen, designed to assist in cell sorting/straining applications.
Catalog No. 50-197-9487 Supplier Chemglass Life Sciences Supplier No. CLS4380009
Strainer Cap Only for FlowTubes, with 35¿m Strainer Mesh, Sterile, 20 bags of 25 Caps. Cap contains a 35¿m nylon mesh screen, designed to assist in cell sorting/straining applications.