Learn More
Thomas Scientific Cardboard Cell Dividers, 36 cell
SDP
Catalog No. NC9817475
Encompass_Preferred
Standard cardboard cell dividers fit all 2" and 3" standard cardboard and metal boxes.
Catalog No. NC9817475 Supplier Thomas Scientific Supplier No. 1149N51
Standard cardboard cell dividers fit all 2" and 3" standard cardboard and metal boxes.