Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex L/S High-Performance Precision Pump Tubing, Gore STA-PURE PFL-Series, L/S 36; 14""/Pk
Catalog No. NC1512539
Encompass
Count on this high-performance pump tubing for challenging applications. Genuine Masterflex tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment. Made by a global market leader in range of formulations for most extensive chemical compatibility, the tubing is precision extruded to tight tolerances for high repeatability, accuracy and reproducibility.
Catalog No. NC1512539 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX9624236
Count on this high-performance pump tubing for challenging applications. Genuine Masterflex tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment. Made by a global market leader in range of formulations for most extensive chemical compatibility, the tubing is precision extruded to tight tolerances for high repeatability, accuracy and reproducibility.