Learn More
Advanced Medical Systems 36 inch Microbore Extension Set, 100/cs
Catalog No. NC9018400
Encompass_Preferred
36 in. Ultra-microbore extension set with slide clamp, male and female luer locks. Approx. priming volume 0.2 mL. Non-DEHP tubing. This device is not made with dry or natural rubber latex. 100 per case.
Catalog No. NC9018400 Supplier Advanced Medical Systems Supplier No. AMS483C
36 in. Ultra-microbore extension set with slide clamp, male and female luer locks. Approx. priming volume 0.2 mL. Non-DEHP tubing. This device is not made with dry or natural rubber latex. 100 per case.