Learn More
SAI Infusion Technologies 36"" Extension Set - Microvolume Tubing, Male and Female Luers, each
SDP
Supplier:  SAI Infusion Technologies EXT361
Encompass
This item has a minimum qty of 15 per supplier requirements.
Microvolume fluid extension line, 36" with male and female luers. SAI extension lines are constructed with non-DEHP PVC tubing. The EXT-36 extension line has one female and one male luer end, perfect for connecting between a pump and luer swivel. Additional sizes are also available. Sold individually, 1/each. Product is for laboratory animal research only. ** If a COA is required it must be requested when the order is placed or a $50.00 fee will be apply.  Contact Thermo Fisher Customer Service at 800-766-7000 to request to COA. **
Catalog No. NC0605217