Learn More
MSC Value Collection Gage Block Set, Steel, 1 ea
Catalog No. NC1416924
Encompass
This gage block set is designed for precision measurement and calibration applications. It includes 36 steel blocks covering a range from 0.05 to 4 in., suitable for general purpose use where economy grade accuracy is sufficient.
  • Rectangular shape
  • Steel material
  • Economy accuracy grade
  • Individually serialized
  • Tolerance of 0.0001 in.; +/- 50 micro in.
  • Step increments: (9) 0.010 in.; (9) 0.0001 in.; (9) 0.001 in.; (5) 0.100 in.
  • Includes a fitted case
  • Meets ASME B89.1.9-2002 standards
Catalog No. NC1416924 Supplier MSC Supplier No. 06461370
This gage block set is designed for precision measurement and calibration applications. It includes 36 steel blocks covering a range from 0.05 to 4 in., suitable for general purpose use where economy grade accuracy is sufficient.
  • Rectangular shape
  • Steel material
  • Economy accuracy grade
  • Individually serialized
  • Tolerance of 0.0001 in.; +/- 50 micro in.
  • Step increments: (9) 0.010 in.; (9) 0.0001 in.; (9) 0.001 in.; (5) 0.100 in.
  • Includes a fitted case
  • Meets ASME B89.1.9-2002 standards