Learn More
Ace Glass, Inc. Rotameter tube, size 36, no stainless steel float supplied, with glass float:0.24-14.3 liter/min air flow, 6.5-300 mL/min water flow
SDP
Catalog No. 50974816
Encompass
Catalog No. 50-974-816 Supplier Ace Glass, Inc. Supplier No. 748125