Learn More
Vici Valco Direct-Connect 360 µm FS Tubing to 1/16" Fitting Detail, Hex main body
SDP
Catalog No. NC1605589
Encompass_Preferred
Fingertight to 25,000 psi (with 150 micron ID tubing). Eliminates dead volume present in competing designs. For use in valves with port size of 150 microns or smaller.
Catalog No. NC1605589 Supplier Vici Valco Supplier No. C360IZR1HNPKG
Fingertight to 25,000 psi (with 150 micron ID tubing). Eliminates dead volume present in competing designs. For use in valves with port size of 150 microns or smaller.