Learn More
MSC 361F CLOTH BELT
Catalog No. 5000802995
Encompass
The Cloth Belt 361F offers durability and consistent performance for grinding, blending, deburring, and finishing various metals. It features durable aluminum oxide abrasive grains and a heat-resistant resin coating, making it suitable for both wet and dry applications in grinding sheets, coils, and centerless operations.
  • Durable aluminum oxide abrasive grains for fast cutting
  • Heat-resistant resin coating for durability under pressure, heat, and water
  • Ideal for grinding sheets and coils, and for centerless and backstand applications
  • Suitable for both wet and dry applications
  • Polyester cloth backing, XF-weight
  • Effective on various materials including stainless steel
Catalog No. 50-008-02995 Supplier MSC Supplier No. 69820082
The Cloth Belt 361F offers durability and consistent performance for grinding, blending, deburring, and finishing various metals. It features durable aluminum oxide abrasive grains and a heat-resistant resin coating, making it suitable for both wet and dry applications in grinding sheets, coils, and centerless operations.
  • Durable aluminum oxide abrasive grains for fast cutting
  • Heat-resistant resin coating for durability under pressure, heat, and water
  • Ideal for grinding sheets and coils, and for centerless and backstand applications
  • Suitable for both wet and dry applications
  • Polyester cloth backing, XF-weight
  • Effective on various materials including stainless steel