Learn More
Electron Microscopy Sciences ProPlate™ 364 Square Well Adhesive SBS Microtiter Plate
SDP
Catalog No. 5034339
Encompass_Preferred
Patented ProPlate technology for high throughput robotic processing of microarrays. Grace BioLabs developed bottomless SBS compliant microtiter plate superstructures for use with robotic processors. Features guaranteed flat SBS compliant microtiter plate format which enables robotic processing of assays. It is compatible with glass or plastic substrates and has a water-tight seal between superstructure and substrate. Adhesive SBS Microtiter Plate. Black Polystyrene.
Catalog No. 50-343-39 Supplier Electron Microscopy Sciences Supplier No. 6348505
Patented ProPlate technology for high throughput robotic processing of microarrays. Grace BioLabs developed bottomless SBS compliant microtiter plate superstructures for use with robotic processors. Features guaranteed flat SBS compliant microtiter plate format which enables robotic processing of assays. It is compatible with glass or plastic substrates and has a water-tight seal between superstructure and substrate. Adhesive SBS Microtiter Plate. Black Polystyrene.