Learn More
Chemglass Life Sciences Chemglass Replacement Bulb, 365 nm - for CLS-1625 Series Handheld Cordless UV Lamp
SDP
Catalog No. NC3235042
Encompass
Replacement 365 nm UV bulb for Chemglass handheld, cordless UV lamps. Sold individually as a service part for the handheld CLS-1625 series; provides the 365 nm wavelength used for fluorescence observation and inspection.
  • 365 nm wavelength.
  • Designed for handheld, cordless UV lamps with safety switches.
  • Compatible with 6 W lamp systems.
  • Sold individually (each).
Catalog No. NC3235042 Supplier Chemglass Life Sciences Supplier No. CLS1625365
Replacement 365 nm UV bulb for Chemglass handheld, cordless UV lamps. Sold individually as a service part for the handheld CLS-1625 series; provides the 365 nm wavelength used for fluorescence observation and inspection.
  • 365 nm wavelength.
  • Designed for handheld, cordless UV lamps with safety switches.
  • Compatible with 6 W lamp systems.
  • Sold individually (each).