Learn More
Medchemexpress LLC HY-W067705 500mg Medchemexpress, 3,6,9-Trioxaundecanedioic Acid CAS:13887-98-4 Purity:>98%
Supplier: Medchemexpress LLC HYW067705500MG
Medchemexpress, HY-W067705 500mg 3,6,9-Trioxaundecanedioic Acid CAS:13887-98-4 369-Trioxaundecanedioic Acid is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.