Learn More
Bellco Glass, Inc. 3.6mL CF Cryogenic Vial Inter
SDP
Supplier:  Bellco Glass, Inc. 229917000
Encompass_Preferred
CELLTREAT Freeze Cryogenic Vials are designed for storing biological material at temperatures as low as 196°C in the gas phase of liquid nitrogen. CELLTREAT product descriptions are based on the highest graduation mark on the vial. Other brands may name vials based on actual liquid capacity. External thread cap features integrated leakproof seal a long skirt for easy one hand aseptic methods and a thread design allowing it to be removed or sealed with a mere 1 1/4 turn. internal thread cap prevents contamination and uses Oring silicone seal to ensure positive seal at any temperature.Specifications Available in round bottom or selfstanding options Selfstanding resealable bags offer a unique storage solution Caps have textured ridges to provide gripping when wearing gloves Contrasting black graduations are large and easy to view Large white marking area for writing also provides backdrop t
Catalog No. 50-232-9340