Learn More
MSC Koch Filter Air Filter, 36 in, MERV 4, 15 ft
Catalog No. 502241161
Encompass
This panel air filter media is 36 in wide with a MERV 4 rating, constructed from polyester material. It is designed for use in applications requiring up to 500 FPM airflow and is LEED certified.
  • Polyester media construction
  • Overall thickness of 1/2 in
  • MERV 4 rating
  • Arrestance efficiency of 0.78%
  • Maximum airflow of 500 FPM
  • Suitable for use with Continental systems
  • ASHRAE 52.2-2012 compliant
  • LEED certified and made from recycled content
This item is not returnable. View return policy
Catalog No. 50-224-1161 Supplier MSC Supplier No. 72999121
This panel air filter media is 36 in wide with a MERV 4 rating, constructed from polyester material. It is designed for use in applications requiring up to 500 FPM airflow and is LEED certified.
  • Polyester media construction
  • Overall thickness of 1/2 in
  • MERV 4 rating
  • Arrestance efficiency of 0.78%
  • Maximum airflow of 500 FPM
  • Suitable for use with Continental systems
  • ASHRAE 52.2-2012 compliant
  • LEED certified and made from recycled content