Learn More
Tewes Corporation 36X48 6MIL FLAT BAG 50/CS
SDP
Catalog No. NC0889481
Encompass_Preferred
Flat Poly Bags with Extra Heavy Duty Protection when Packaging sharp or very heavy items that may cause tears or puntures; Strong enough to reuse multiple times; Meet FDA and USDA specifications; Bags can be heat sealed, tied, stapled or taped to close. 50/CASE.
Catalog No. NC0889481 Supplier Tewes Corporation Supplier No. F36486
Flat Poly Bags with Extra Heavy Duty Protection when Packaging sharp or very heavy items that may cause tears or puntures; Strong enough to reuse multiple times; Meet FDA and USDA specifications; Bags can be heat sealed, tied, stapled or taped to close. 50/CASE.