Learn More
Azer Scientific 37% Formaldehyde
SDP
Catalog No. 21402492
Change view
Click to view available options
Quantity:
4 x 1 gal.
Catalog No. Quantity
21-402-492 4 x 1 gal.
1 options
Catalog No. 21-402-492 Supplier Azer Scientific Supplier No. F371G
Add to Cart
Add to Cart
Intended for use in the most critical applications in microscopy and histology
  • Azer Scientific 37% formaldehyde comes in multiple sizes including 32oz.,1 Gallon, 5 Gallon and 55 Gallon containers
  • It is intended for use in the most critical applications in microscopy and histology
  • 37% formaldehyde can be used with automated or manual protocols and has a long, stable shelf life
  • Our formula is stabilized with methanol
  • Our 5 gallon cube features a spigot for easy use
  • Stabilized with methanol
  • Used in the most critical applications in microscopy and histology
  • Long, stable shelf life
Specifications
Quantity 4 x 1 gal.