Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ 3730/3730xl DNA Analyzer Sequencing Standards, BigDye™ Terminator v1.1

Catalog No. 4336799
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-367-99 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 43-367-99 Supplier Applied Biosystems™ Supplier No. 4336799
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Kit contains 4 tubes of hsp69 standard sequence used for instrument and spectral calibrations

This material is used with the 3730, and 3730xl DNA Analyzers.

  • Kit contains 4 tubes of hsp69 standard sequence used for instrument and spectral calibrations
  • This material is used with the 3730, and 3730xl DNA Analyzers.
  • Store kit at -15 to -25°C
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage This kit contains 4 tubes of hsp69 standard sequence used for instrument and spectral calibrations. This material is used with the 3730, and 3730xl DNA Analyzers. Store kit at -15 to -25°C.
For Use With (Equipment) 3730xl DNA Analyzer, 3730 DNA Analyzer
For Use With (Application) Sequencing
Label or Dye ROX, TAMRA™, Rhodamine 110, Rhodamine 6G
Product Line BigDye Terminator, BigDye
Product Type Sequencing Standards
Quantity 1 Kit
Shipping Condition Dry Ice
I see another sequence running under mine. What can cause this?

Some of the causes of another sequence appearing under the primary sequence are:

• Contamination: There is more than one species of DNA present (e.g., multiple PCR products).

• Primers: Primer is annealing in more than one location on the template, primer dimer, primer degradation or not manufactured properly resulting in N+1 or N-1, carry over from PCR reaction, or primers pairs in a multiplex reaction that are not appropriate for multiplexing (i.e., primers anneal inappropriately).

• Spectral/Matrix: If the raw data signal intensity of the sample is too high or saturated it can exceed the amount of color bleedthrough (or spectral overlap) that the matrix (310) or spectral (3130, 3730, 3500) are removing, resulting in secondary peaks appearing in a very specific pattern (e.g., a red peak always appearing under a green peak). A change in the camera, laser, or optical alignment requires that a new matrix be made or a new spectral calibration be performed.

For more information on more than one sequence or set of peaks in a sequencing run, please refer to the DNA Sequencing by Capillary Electrophoresis: Applied Biosystems Chemistry Guide: Second Edition (Cat. No. 4305080, Rev. C). The guide can be found by searching the Thermo Fisher Scientific website with the catalog number 4305080.

How often should a spectral calibration be run on the Applied Biosystems 3730 Series DNA Analyzer?

There is no fixed time as to when a spectral must be run. However, some users prefer to run a new spectral at a preset time to ensure it is up to date. Data quality will be the largest indicator of when a new spectral is required. A separate spectral calibration needs to be performed for each dye set, array type, and array length. For G5-RCT dye set users only: a spectral calibration must be re-run if the array detection cell is moved/repositioned. Refer to FAQ "When should I run a spectral calibration?" for more information on when a spectral should be run.

When should I run a spectral calibration on the Applied Biosystems 3730 Series DNA Analyzer?

A new spectral calibration should be run:

- Whenever a new dye set is used on an instrument.
- After any optics adjustment performed by a service engineer.
- If the instrument has been moved.
- If you see a decrease in spectral separation (pull-up and/or pull-down peaks).
- If you alter any conditions (dye set, array type, or array length).
- For G5-RCT, Any4dye-HDR, and S dye set users only: Any time the array detection cell is moved/repositioned.

What is a spectral calibration?

A spectral calibration is an algorithm applied to raw data, which converts it into the component 4 or 5 dye data stored in the sample files. A spectral is created for a specific dye set (combination of dyes), array type (4 or 16 capillaries), and array length (36cm or 50cm). It is used to correct for the natural overlap of the fluorescent dyes.

What is the shelf life of the sequencing standard?

The sequencing standard has a shelf life of one year from the date of shipment, unless otherwise listed on the CofA. Our Terms and Conditions of sale can be found here(https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf).

Can I re-inject sequencing standard that is already been resuspended in Hi‑Di formamide? If so, how do I store it between uses? How long will it last?

The sequencing standard is intended to be used after resuspension in Hi‑Di Formamide. It can be injected multiple times; the number of times will vary depending on the instrument-specific standard used. This information can be found in the product insert. The sequencing standard should be stable for 24 hours on the instrument.


For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.