Learn More
Accuflow Systems Saint-Gobain C-Flex Tubing, 3/8 in ID, Translucent, 50 ft
Catalog No. 502365918
Encompass_Preferred
C-Flex TPE tubing, formulation 374, is a translucent thermoplastic elastomer tubing supplied as a 50 ft coil. It is designed for high-purity fluid transfer where sealability and weldability are required, and is commonly used in biopharmaceutical and pharmaceutical process systems.
  • Thermoplastic elastomer construction for flexibility and chemical compatibility.
  • Translucent appearance for visual flow monitoring.
  • Inner diameter: 3/8 in, outer diameter: 1/2 in.
  • Supplied as a 50 ft coil for continuous runs.
  • Compatible with aseptic welding and sealing processes.
  • Intended for biopharmaceutical and high-purity fluid-transfer applications.
Catalog No. 50-236-5918 Supplier Accuflow Systems Supplier No. 3743752
C-Flex TPE tubing, formulation 374, is a translucent thermoplastic elastomer tubing supplied as a 50 ft coil. It is designed for high-purity fluid transfer where sealability and weldability are required, and is commonly used in biopharmaceutical and pharmaceutical process systems.
  • Thermoplastic elastomer construction for flexibility and chemical compatibility.
  • Translucent appearance for visual flow monitoring.
  • Inner diameter: 3/8 in, outer diameter: 1/2 in.
  • Supplied as a 50 ft coil for continuous runs.
  • Compatible with aseptic welding and sealing processes.
  • Intended for biopharmaceutical and high-purity fluid-transfer applications.