Learn More
Accuflow Systems Saint-Gobain C-Flex Tubing, TPE, 3/8 x 5/8 in, 150 ft
Catalog No. NC2312793
Encompass_Preferred
Translucent thermoplastic elastomer (TPE) tubing designed for biopharmaceutical and laboratory fluid transfer applications. Supplied on a compact mini spool for convenient handling, this tubing is suitable for peristaltic pumping, aseptic welding, and sterilization workflows.
  • Thermoplastic elastomer tubing, translucent and flexible.
  • Inner diameter 3/8 in, outer diameter 5/8 in.
  • 150 ft mini spool for compact storage and deployment.
  • Sealable and weldable for aseptic connections.
  • Compatible with gamma irradiation and autoclave sterilization.
  • Suitable for peristaltic pumps, with lower gas permeability than silicone.
  • Operating temperature range -45 °C to 135 °C.
Catalog No. NC2312793 Supplier Accuflow Systems Supplier No. 3743754M
Translucent thermoplastic elastomer (TPE) tubing designed for biopharmaceutical and laboratory fluid transfer applications. Supplied on a compact mini spool for convenient handling, this tubing is suitable for peristaltic pumping, aseptic welding, and sterilization workflows.
  • Thermoplastic elastomer tubing, translucent and flexible.
  • Inner diameter 3/8 in, outer diameter 5/8 in.
  • 150 ft mini spool for compact storage and deployment.
  • Sealable and weldable for aseptic connections.
  • Compatible with gamma irradiation and autoclave sterilization.
  • Suitable for peristaltic pumps, with lower gas permeability than silicone.
  • Operating temperature range -45 °C to 135 °C.