Learn More
Waters Corp Performance Maintenance Kit, For 3767 Sample Manager, Annual
Catalog No. 502391459 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The kit contains the necessary parts to keep the 3767 Sample Manager instrument operating at peak performance on an annual basis. It includes a vented probe/needle, lubrication kit, injector port fittings, Rheodyne analytical injector rebuild kit, and Rheodyne prep injector rebuild kit. Syringes are recommended for annual replacement but must be purchased separately as they are customer configurable.
  • Contains parts necessary for annual instrument maintenance
  • Designed to maintain peak instrument performance
  • Includes vented probe/needle
  • Includes lubrication kit
  • Includes injector port fittings
  • Includes analytical injector rebuild kit
  • Includes prep injector rebuild kit
Catalog No. 50-239-1459 Supplier Waters Corp Supplier No. 201000333
The kit contains the necessary parts to keep the 3767 Sample Manager instrument operating at peak performance on an annual basis. It includes a vented probe/needle, lubrication kit, injector port fittings, Rheodyne analytical injector rebuild kit, and Rheodyne prep injector rebuild kit. Syringes are recommended for annual replacement but must be purchased separately as they are customer configurable.
  • Contains parts necessary for annual instrument maintenance
  • Designed to maintain peak instrument performance
  • Includes vented probe/needle
  • Includes lubrication kit
  • Includes injector port fittings
  • Includes analytical injector rebuild kit
  • Includes prep injector rebuild kit