Learn More
Waters Corp Performance Maintenance Kit, HPLC, Annual, for 3777C Sample Manager
Catalog No. 502391190 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Performance Maintenance Kit for the 3777C Sample Manager contains all of the Quality Parts necessary to keep your instrument operating at peak performance on an annual basis.
  • Lubrication kit
  • Injector seal
  • 3 psi Check Valve
  • Injector waste tube
  • 3 Replacement needles
  • 10μL Sample Loop
  • Holding loop
  • 100μL Syringe
Catalog No. 50-239-1190 Supplier Waters Corp Supplier No. 201000318
The Performance Maintenance Kit for the 3777C Sample Manager contains all of the Quality Parts necessary to keep your instrument operating at peak performance on an annual basis.
  • Lubrication kit
  • Injector seal
  • 3 psi Check Valve
  • Injector waste tube
  • 3 Replacement needles
  • 10μL Sample Loop
  • Holding loop
  • 100μL Syringe