Learn More
McMaster-Carr 37 Degree Flared Fitting for Steel Tubing
Catalog No. NC1457070
Encompass
37 Degree Flared Fitting for Steel TubingMale Adapter for 1/4" Tube OD x 1/4 BSPT, 4500 PSI
Catalog No. NC1457070 Supplier McMaster-Carr Supplier No. 4545K163
37 Degree Flared Fitting for Steel TubingMale Adapter for 1/4" Tube OD x 1/4 BSPT, 4500 PSI