Learn More
Electron Microscopy Sciences With Immersion Oil Type LDF extremely low fluorescence is achieved. Possesses the highest resolution of the oils for fluorescence applications. Replacement for type DF.
SDP
Catalog No. NC3552920
Encompass_Preferred
With Immersion Oil Type LDF extremely low fluorescence is achieved. Possesses the highest resolution of the oils for fluorescence applications. Replacement for type DF.
Catalog No. NC3552920 Supplier Electron Microscopy Sciences Supplier No. 1691912
With Immersion Oil Type LDF extremely low fluorescence is achieved. Possesses the highest resolution of the oils for fluorescence applications. Replacement for type DF.