Learn More
Electron Microscopy Sciences 37mm Air Sampling Cassette MCE Filters with 0.45 µm pores, 3 Piece housing
SDP
Supplier:  Electron Microscopy Sciences 6521845
Encompass
The 37 mm Air Sampling Cassette w/MCE Filters are designed to meet all NIOSH, OSHA, and EPA requirements for air sampling standards. All 37 mm cassette housings are manufactured in house and are available in both two and three piece configurations. All 37 mm cassettes are individually assembled and preloaded under stringent quality assurance standards with only the finest quality membrane and support pads. Suitable for lead sampling.
Pores: 0.45 µm
Housing: 3 Piece
Catalog No. 50-338-58