Learn More
Electron Microscopy Sciences 37 mm Air Sampling Cassette PTFE Filters with 2.0 µm pores, 2 Piece housing
SDP
Supplier:  Electron Microscopy Sciences 6522223
Encompass
The 37 mm Air Sampling Cassette w/"PTFE" Filters are designed to meet all NIOSH, OSHA, and EPA requirement for air sampling standards. All 37 mm cassette housings are manufactured in house and are available in both two and three piece configurations. All 37 mm cassettes are individually assembled and preloaded under stringent quality assurance standards with only the finest quality membrane and support pads. With support (Zefluor).
Pores: 2.0 µm
Housing: 2 Piece
Catalog No. 50-338-72
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.