Learn More
Electron Microscopy Sciences 37mm Air Sampling Cassette MCE Filters with 5.0 µm pores, 2 Piece housing
SDP
Supplier:  Electron Microscopy Sciences 6521925
Encompass
The 37 mm Air Sampling Cassette w/PVC Filters meet all NIOSH, OSHA, and EPA requirement for air sampling standards. All 37 mm cassette housings are manufactured in house and are available in both two and three piece configurations and are individually assembled and preloaded under stringent quality assurance standards with only the finest quality membrane and support pads. Preloaded and ready-to-use. Leak-free design.
Pores: 5.0 µm
Housing: 2 Piece
Catalog No. 50-338-59