Learn More
Ace Glass, Inc. 37mm quartz graded seals for connecting Pyrex or Kimax borosilicate glass to Quartz glass. Quartz length is 7.6cm, and borosilicate length is 10.2cm.
SDP
Catalog No. 50974606
Encompass
37mm quartz graded seals for connecting Pyrex or Kimax borosilicate glass to Quartz glass. Quartz length is 7.6cm, and borosilicate length is 10.2cm.
Catalog No. 50-974-606 Supplier Ace Glass, Inc. Supplier No. 845546
Add to Cart
Add to Cart
37mm quartz graded seals for connecting Pyrex or Kimax borosilicate glass to Quartz glass. Quartz length is 7.6cm, and borosilicate length is 10.2cm.