Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences WHEATON™ 38-400 Black Phenolic Cap, PE Cone Liner
SDP

Catalog No. 02927140 Shop All DWK Life Sciences Products
Catalog No. 02-927-140 Supplier DWK Life Sciences Supplier No. W240541
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

WHEATON™ 38-400 Black Phenolic Cap, PE Cone Liner provides a secure closure for WHEATON™ Valumetric™ graduated bottles. Features a polyethylene cone liner for a reliable seal. Available in cases of 576 caps (48 per pack) with a 38-400 thread finish.

  • Replacement caps for Wheaton Valumetric™ graduated bottles with screw cap tops
  • Cap size: 38-400 thread finish (GSI 400 = 1 turn)
  • Opening outer diameter including threads: 38mm
  • Liner material: Polyethylene (PE) cone liner
  • Pack quantity: 48 caps per pack; 576 caps per case
TRUSTED_SUSTAINABILITY

Specifications

Color Black
Material Phenolic Resin
Quantity 576/Case
Sterility Non-sterile
Liner Material Polyethylene
Neck Finish 38-400
For Use With (Equipment) Wheaton screw cap bottles/vials
Autoclavable Not Autoclavable
Disposable Reusable
Product Type Cone-lined Cap
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.