Learn More
Thermco Products Inc -38/50C X 1.0C
Catalog No. NC1284530
Encompass_Preferred
ASTM S5C thermometer (blue spirit) certified at the ASTM recommended temperatures and calibrated against N.I.S.T. standards. A N.I.S.T. traceable document is completed in the Thermco ISO17025/ILAC accedited laboratory.
Catalog No. NC1284530 Supplier Thermco Products Inc Supplier No. ACC005CBLS
ASTM S5C thermometer (blue spirit) certified at the ASTM recommended temperatures and calibrated against N.I.S.T. standards. A N.I.S.T. traceable document is completed in the Thermco ISO17025/ILAC accedited laboratory.