Learn More
Beckman Coulter 38.5 mL, Sterile + Certified Free Open-Top Thinwall Ultra-Clear Tube, 25 x 89mm, 48/pk
Catalog No. NC2010547
Encompass_Preferred
This round bottom tube is designed for use in Beckman Coulter instruments and rotors. Ultra-Clear tubes are transparent, allowing for easy location of visibly banded or pelleted samples.
Catalog No. NC2010547 Supplier Beckman Coulter Supplier No. C14292
This round bottom tube is designed for use in Beckman Coulter instruments and rotors. Ultra-Clear tubes are transparent, allowing for easy location of visibly banded or pelleted samples.