Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 38-azido-3,6,9,12,15,18,21,24,27,30,33,36-dodecaoxaoctatriacontan-1-amine | 2803478-25-1 | 98.0% | 614.73 | C26H54N4O12 | 100 MG
SDP

Catalog No. 5000284408
Encompass_Preferred

Azido-PEG12-amine is an azide-terminated polyethylene glycol (PEG12) linker bearing a terminal primary amine used in bioconjugation and click-chemistry applications.

  • Azide group enables click chemistry (azide-alkyne cycloaddition).
  • Terminal primary amine reacts with carboxylic acids, activated NHS esters, and carbonyls.
  • PEG12 spacer increases aqueous solubility and reduces steric hindrance.
  • Molecular weight 614.73; formula C26H54N4O12.
  • High purity (98.0% by NMR) with 1H NMR and MS consistent with structure.
  • Storage: sealed, away from moisture and light; in solvent store at -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-84408 Supplier Medchemexpress LLC Supplier No. HYW590569100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Azido-PEG12-amine is an azide-terminated polyethylene glycol (PEG12) linker bearing a terminal primary amine used in bioconjugation and click-chemistry applications.

  • Azide group enables click chemistry (azide-alkyne cycloaddition).
  • Terminal primary amine reacts with carboxylic acids, activated NHS esters, and carbonyls.
  • PEG12 spacer increases aqueous solubility and reduces steric hindrance.
  • Molecular weight 614.73; formula C26H54N4O12.
  • High purity (98.0% by NMR) with 1H NMR and MS consistent with structure.
  • Storage: sealed, away from moisture and light; in solvent store at -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.