Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 38-azido-3,6,9,12,15,18,21,24,27,30,33,36-dodecaoxaoctatriacontan-1-amine | 2803478-25-1 | 98.0% | 614.73 g/mol | C26H54N4O12 | 25 MG
SDP

Catalog No. 5000284404
Encompass_Preferred

Azido-PEG12-amine is a PEG-based bifunctional linker with an azide at one terminus and a primary amine at the other. It functions as a water-soluble spacer (approximately 12 ethylene glycol units) used in click chemistry, bioconjugation, PROTAC and ADC linker design.

  • Azide and primary amine functional groups enable click chemistry and amide coupling.
  • Approximately 12 ethylene glycol units provide flexible, hydrophilic spacing.
  • Molecular weight 614.73 g/mol for predictable linker length and mass.
  • Purity 98.0% (NMR) as supplied in certificate of analysis.
  • Colorless to light yellow liquid physical form for ease of handling.
  • Stable when stored sealed and protected from moisture and light; in solvent: -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-84404 Supplier Medchemexpress LLC Supplier No. HYW59056925MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Azido-PEG12-amine is a PEG-based bifunctional linker with an azide at one terminus and a primary amine at the other. It functions as a water-soluble spacer (approximately 12 ethylene glycol units) used in click chemistry, bioconjugation, PROTAC and ADC linker design.

  • Azide and primary amine functional groups enable click chemistry and amide coupling.
  • Approximately 12 ethylene glycol units provide flexible, hydrophilic spacing.
  • Molecular weight 614.73 g/mol for predictable linker length and mass.
  • Purity 98.0% (NMR) as supplied in certificate of analysis.
  • Colorless to light yellow liquid physical form for ease of handling.
  • Stable when stored sealed and protected from moisture and light; in solvent: -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.