Learn More
Noir Lasers #38 SHIELD ARGON 532NM LASERS
SDP
Catalog No. 501050537
Encompass_Preferred
NoIR LaserShields, #38 - large fitover frame with side shields, ANSI Specifications 190-532nm, 7+OD, CE Certified, polycarbonate lens, 49% VLT, CE Certified/ANSI Certified. Certifications available upon request. Hardshell Case with cleaning cloth and strap
This item is not returnable. View return policy
Catalog No. 50-105-0537 Supplier Noir Lasers Supplier No. ARG#38
NoIR LaserShields, #38 - large fitover frame with side shields, ANSI Specifications 190-532nm, 7+OD, CE Certified, polycarbonate lens, 49% VLT, CE Certified/ANSI Certified. Certifications available upon request. Hardshell Case with cleaning cloth and strap