Learn More
Chemglass Life Sciences Stopper, Safe-Lab, Flask Length, PTFE, Size 38
SDP
Catalog No. CG301908
Encompass_Preferred
Stopper, Safe-Lab, Flask Length, PTFE, Size 38. Flask length PTFE stopper has a Polypropylene extraction nut permitting easy removal of the stopper from the flask. All sizes are solid.
Catalog No. CG301908 Supplier Chemglass Life Sciences Supplier No. CG301908
Stopper, Safe-Lab, Flask Length, PTFE, Size 38. Flask length PTFE stopper has a Polypropylene extraction nut permitting easy removal of the stopper from the flask. All sizes are solid.