Learn More
Chemglass Life Sciences Vacuum Trap, 38mm Body OD x 250mm Body Length, 18mm OD Side and Inner Tubes
SDP
Supplier:  Chemglass Life Sciences CG451204
Encompass_Preferred
Vacuum Trap, 38mm Body OD x 250mm Body Length, 18mm OD Side and Inner Tubes. Similar to CG-4510 but with top inlet tube bent at a 90º angle.
Catalog No. CG451204