Learn More
Molecular Devices Molecular Devices Microplate Wash Head, 384-Well, AquaMax 2000/4000 Compatible
Catalog No. 5000014706
Encompass_Preferred
Replacement 384-well microplate wash head designed for use with AquaMax 2000 and AquaMax 4000 microplate washers. It provides a 384-well configuration for automated plate washing in biochemical and cell-based assays and is interchangeable with other compatible wash heads.
  • Designed for 384-well microplate formats.
  • Enables automated plate washing for biochemical and cell-based assays.
  • Interchangeable with compatible microplate washer head systems.
  • Supports gentle cell wash protocols when used with appropriate wash settings.
  • Facilitates high-throughput processing and reduces manual handling.
Catalog No. 50-000-14706 Supplier Molecular Devices Supplier No. 03105219T
Add to Cart
Add to Cart
Request a Quote
Replacement 384-well microplate wash head designed for use with AquaMax 2000 and AquaMax 4000 microplate washers. It provides a 384-well configuration for automated plate washing in biochemical and cell-based assays and is interchangeable with other compatible wash heads.
  • Designed for 384-well microplate formats.
  • Enables automated plate washing for biochemical and cell-based assays.
  • Interchangeable with compatible microplate washer head systems.
  • Supports gentle cell wash protocols when used with appropriate wash settings.
  • Facilitates high-throughput processing and reduces manual handling.